Skip to Content
Merck
All Photos(4)

Documents

T3203

Sigma-Aldrich

p-Terphenyl

≥99.5% (HPLC)

Synonym(s):

1,4-Diphenylbenzene, PT, PTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4C6H5
CAS Number:
Molecular Weight:
230.30
Beilstein:
1908447
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

7.95 (vs air)

Assay

≥99.5% (HPLC)

bp

389 °C (lit.)

mp

212-213 °C (lit.)

SMILES string

c1ccc(cc1)-c2ccc(cc2)-c3ccccc3

InChI

1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H

InChI key

XJKSTNDFUHDPQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hengde Li et al.
Genetica, 143(3), 299-304 (2015-02-07)
Genetic prediction of quantitative traits is a critical task in plant and animal breeding. Genomic selection is an accurate and efficient method of estimating genetic merits by using high-density genome-wide single nucleotide polymorphisms (SNP). In the framework of linear mixed
Aurélia Caputo et al.
Biology direct, 10, 5-5 (2015-04-19)
Alterations in gut microbiota composition under antibiotic pressure have been widely studied, revealing a restricted diversity of gut flora, including colonization by organisms such as Enterococci, while their impact on bacterial load is variable. High-level colonization by Akkermansia muciniphila, ranging
Renate B Schnabel et al.
Lancet (London, England), 386(9989), 154-162 (2015-05-12)
Comprehensive long-term data on atrial fibrillation trends in men and women are scant. We aimed to provide such data through analysis of the Framingham cohort over 50 years. We investigated trends in incidence, prevalence, and risk factors for atrial fibrillation
Romain Duval et al.
Scientific reports, 5, 10750-10750 (2015-06-05)
Protein tyrosine phosphatases (PTPs) are involved in numerous signaling pathways and dysfunctions of certain of these enzymes have been linked to several human diseases including cancer and autoimmune diseases. PTPN2 is a PTP mainly expressed in hematopoietic cells and involved
Thomas A Lanz et al.
PloS one, 10(3), e0121744-e0121744 (2015-03-19)
Increased protein levels of striatal-enriched tyrosine phosphatase (STEP) have recently been reported in postmortem schizophrenic cortex. The present study sought to replicate this finding in a separate cohort of postmortem samples and to extend observations to striatum, including subjects with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service