SYNF580050
Synthware™ storage tube with high vacuum valve
capacity 50 mL, valve size 0-4 mm, tube O.D. × L 28 mm × 120 mm
Synonym(s):
Synthware F580050, Schlenk tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
black cap
cap (plug seal)
colorless tube
glass tube
round bottom
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F580050
capacity
50 mL
tube O.D. × L
28 mm × 120 mm
valve size
0-4 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper (“umbrella”) to protect Viton® O-ring on shaft.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service