SYNC144300
Synthware™ Graham condenser
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C179200, distillation condenser, Inland Revenue condenser
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C144300
hose size
10 mm
Related Categories
General description
Synthware™ Graham condenser features a 300 mm jacketed length for superior cooling efficiency. With standardized 24/40 upper and lower joints, it easily fits various laboratory glassware. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)