SLW3152/12A
Pyrex® Volumetric flask, heavy duty, class A, amber, USP/ISO/DIN tolerances,
capacity 100 mL
Synonym(s):
Pyrex, SciLabware 3152/12A
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass flask
Joint
joint ST/NS neck14/23
feature
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
packaging
pkg of 5 ea
manufacturer/tradename
SciLabware 3152/12A
Overall H
170 mm
capacity
100 mL
General description
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
- Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
- Each flask is laser etched with an individual serial number, batch code and date of manufacturer
Features and Benefits
• Complies with ISO 1042 and DIN 12664• Manufactured from borosilicate glass 3.3• Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards• With an amber coating on exterior surfaces to facilitate use with light sensitive solutions• Inscriptions in white enamel• Each flask is laser etched with an individual serial number batch code and date of manufacture• Batch certificate available to download online.
Legal Information
PYREX is a registered trademark of Corning, Inc.