Skip to Content
Merck
All Photos(2)

Key Documents

P9906

Sigma-Aldrich

Poly(ethylene glycol) bis(amine)

average MN 3,400, cross-linking reagent carboxyl reactive, amine

Synonym(s):

Polyethylene glycol, O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) bis(amine), average Mn 3400

form

powder

Quality Level

mol wt

average Mn 3400

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

Ω-end

amine

α-end

amine

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) bis(amine) may be used in the preparation of folic acid-PEG-NH2 conjugate. PEG-bis(amine) has been reported to be used as a component in the making of Au-Fe2O3 NPs based multifunctional nanoprobe for multimodal and ultrasensitive detection of cancer cells.

Preparation Note

Prepared from PEG with an average molecular weight of 3,350

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and characterization of folate targeted nanoparticle drug delivery system
mittal V, et al.
International Journal of Pharmacy and Biological Sciences, V1(2) (2010)
Jian Liu et al.
Chemical communications (Cambridge, England), 49(43), 4938-4940 (2013-04-23)
A multifunctional nanoprobe, which can be used for dual modal imaging and the detection of cancer cells, has been reported.
Evaluation of receptor mediated endocytosis on cellular internalization: a comparative study of pegylated nanoparticles and folate anchored pegylated nanoparticles on mda-mb-231 cells
Mahalwar A, et al.,
International Journal of Biological & Pharmaceutical Research null
Development and characterization of folate targeted nanoparticle drug delivery system
Mittal V, et al.
International Journal of Pharmacy and Biological Sciences, V1(2) (2000)
Stephen J Connon et al.
Bioorganic & medicinal chemistry letters, 12(14), 1873-1876 (2002-06-28)
The synthesis and olefin metathesis activity in protic solvents of 7, a phosphine-free ruthenium alkylidene bound to a hydrophilic solid support are reported. This heterogeneous catalyst promotes relatively efficient ring closing- and cross-metathesis reactions in both methanol and water. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service