Skip to Content
Merck
All Photos(3)

Key Documents

P8010

Sigma-Aldrich

Sodium pyrophosphate tetrabasic

≥95%

Synonym(s):

Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7
CAS Number:
Molecular Weight:
265.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

Quality Level

Assay

≥95%

form

powder

density

2.53 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2/c;;;;1-8(2,3)7-9(4,5)6/h;;;;(H2,1,2,3)(H2,4,5,6)/q4*+1;/p-4

InChI key

FQENQNTWSFEDLI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pyrophosphate tetrabasic (Na4P2O7) is a non-toxic and biocompatible compound used as an electroactive media for exfoliation of the surface coating. It is also be used as an additive in the food industry.

Application

Na4P2O7 can be used as an inorganic additive to improve the stability and electrochemical performance of redox flow batteries.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of inorganic additive sodium pyrophosphate tetrabasic on positive electrolytes for a vanadium redox flow battery
Park S, et al.
Electrochimica Acta, 121(12), 321-327 (2014)
Effects of phosphate additives on the stability of positive electrolytes for vanadium flow batteries
Ding C, et al.
Electrochimica Acta, 164(12), 307-314 (2015)
Wei Qiu et al.
Journal of lipid research, 50(9), 1814-1823 (2009-04-23)
Glucosamine impairs hepatic apolipoprotein B100 (apoB100) production by inducing endoplasmic reticulum (ER) stress and enhancing cotranslational and posttranslational apoB100 degradation (Qiu, W., R. K. Avramoglu, A. C. Rutledge, J. Tsai, and K. Adeli. Mechanisms of glucosamine-induced suppression of the hepatic
Effect of inorganic additive sodium pyrophosphate tetrabasic on positive electrolytes for a vanadium redox flow battery.
Park SK, et al.
Electrochimica Acta, 121, 321-327 (2014)
Electrochemical exfoliation of graphite for producing graphene using saccharin
MK PK, et al.
Royal Society of Chemistry Advances, 5(66), 53865-53869 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service