Skip to Content
Merck
All Photos(1)

Key Documents

P50706

Sigma-Aldrich

1,3-Propanesultone

98%

Synonym(s):

3-Hydroxypropanesulfonic acid γ-sultone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3S
CAS Number:
Molecular Weight:
122.14
Beilstein:
109782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

bp

180 °C/30 mmHg (lit.)

mp

30-33 °C (lit.)

density

1.392 g/mL at 25 °C (lit.)

SMILES string

O=S1(OCCC1)=O

InChI

1S/C3H6O3S/c4-7(5)3-1-2-6-7/h1-3H2

InChI key

FSSPGSAQUIYDCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Propanesultone is a cyclic sulfonic ester mainly used to introduce a propane sulfonic functionality into the organic structure.

Application

1,3-Propanesultone can be used to synthesize:
  • A sulfonic acid functionalized acidic ionic liquid modified silica catalyst that can be used in hydrolysis of cellulose.
  • Zwitterionic-type molten salts with unique ion conductive properties.
  • Zwitterionic organofunctional silicones by the quaternization of organic amine functional silicones.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

340.7 °F

Flash Point(C)

171.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface functionalized ZnO particles designed for the use in transparent nanocomposites.
Khrenov V
Macromolecular Chemistry and Physics, 206(1), 95-101 (2005)
Synthesis of a sulfonic acid functionalized acidic ionic liquid modified silica catalyst and applications in the hydrolysis of cellulose.
Amarasekara AS
Catalysis Communications, 11(13), 1072-1075 (2010)
Synthesis and characterization of zwitterionic silicone sulfobetaine surfactants.
Snow SA
Langmuir, 6(2), 385-391 (1990)
Dong Chan Seo et al.
Polymers, 12(6) (2020-06-24)
To improve both the mechanical and chemical durability of Nafion membranes for polymer electrolyte membrane fuel-cells (PEMFCs), Nafion composite membranes containing sulfonated graphene oxide (SGO) and cerium oxide (CeO2; ceria) were prepared by solution casting. The structure and chemical composition
Additives for High Performance Applications: Chemistry and Applications, 86-86 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service