Skip to Content
Merck
All Photos(3)

Key Documents

P2066

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)

lactide:glycolide 65:35, Mw 40,000-75,000

Synonym(s):

Lactel® B6001-1, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

Quality Level

feed ratio

lactide:glycolide 65:35

mol wt

Mw 40,000-75,000

storage temp.

−20°C

SMILES string

OCC(O)=O.CC(O)C(O)=O

InChI

1S/C3H6O3.C2H4O3/c1-2(4)3(5)6;3-1-2(4)5/h2,4H,1H3,(H,5,6);3H,1H2,(H,4,5)

InChI key

XBBVURRQGJPTHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Inhaled Macrophage Apoptotic Bodies-Engineered Microparticle Enabling Construction of Pro-Regenerative Microenvironment to Fight Hypoxic Lung Injury in Mice.: This study demonstrates the use of Poly(ᴅ,ʟ-lactide-co-glycolide) (PLGA) in engineering microparticles that promote lung tissue regeneration in hypoxic conditions, highlighting its potential in developing advanced therapeutic strategies for respiratory injuries (Liu et al., 2024).

  • Engineered nanoparticles promote cardiac tropism of AAV vectors.: The research explores the role of PLGA nanoparticles in enhancing the targeting efficiency of AAV vectors to cardiac tissues, showcasing its application in gene therapy and cardiovascular disease treatment (Switala et al., 2024).

  • Drug-loaded adhesive microparticles for biofilm prevention on oral surfaces.: This article reports the development of drug-loaded PLGA microparticles designed to prevent biofilm formation on oral surfaces, demonstrating the polymer′s utility in dental applications and biofilm-associated infections (Oh et al., 2024).

  • Dual-layer conduit containing VEGF-A - Transfected Schwann cells promotes peripheral nerve regeneration via angiogenesis.: PLGA-based dual-layer conduits are utilized to support nerve regeneration by enhancing angiogenesis, indicating its importance in neuroregenerative medicine and peripheral nerve injury treatments (Huang et al., 2024).

  • Application of Doxorubicin-loaded PLGA nanoparticles targeting both tumor cells and cancer-associated fibroblasts on 3D human skin equivalents mimicking melanoma and cutaneous squamous cell carcinoma.: The study highlights the dual-targeting capability of PLGA nanoparticles in delivering chemotherapy drugs to both tumor cells and the surrounding supportive stroma, emphasizing its role in advanced cancer therapies (He et al., 2024).

Features and Benefits

Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.

Physical form

Biocompatible, biodegradable polymer.

Legal Information

Lactel is a registered trademark of Evonik Corp

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleftheria Babaliari et al.
International journal of molecular sciences, 19(7) (2018-07-18)
Cell responses depend on the stimuli received by the surrounding extracellular environment, which provides the cues required for adhesion, orientation, proliferation, and differentiation at the micro and the nano scales. In this study, discontinuous microcones on silicon (Si) and continuous
Shu-Chun Chuang et al.
Parasites & vectors, 6, 34-34 (2013-02-13)
Current development efforts of subunit vaccines against Toxoplasma gondii, the etiological agent of toxoplasmosis, have been focused mainly on tachyzoite surface antigen 1 (SAG1). Since microparticles made from poly (lactide-co-glycolide) (PLG) polymers have been developed as safe, potent adjuvants or
Rongcai Liang et al.
International journal of pharmaceutics, 454(1), 344-353 (2013-07-23)
Peptide or protein degradation often occurs when water flows into the dosage form. The aim of this study was to investigate the effect of water on exenatide acylation in poly(lactide-co-glycolide) (PLGA) microspheres. Exenatide-loaded PLGA microspheres were incubated at different relative
Teresa Musumeci et al.
International journal of pharmaceutics, 440(2), 135-140 (2012-10-20)
Melatonin, a neurohormone secreted by the pineal gland, is able to modulate intraocular pressure (IOP). The aim of this study was to generate nanoparticle (NPs) sustained release formulations that allow to extend the pre-corneal residence time of melatonin, thus prolonging
Igor Jeroukhimov et al.
Journal of the American College of Surgeons, 218(1), 102-107 (2013-11-12)
Chronic pain after inguinal hernia repair occurs in 16% to 62% of patients. The underlying mechanism probably involves sensory nerve damage and abnormal healing that might be influenced by the materials chosen for the procedure. We hypothesize that nonabsorbable sutures

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Protocols

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service