Skip to Content
Merck
All Photos(3)

Documents

O9409

Sigma-Aldrich

2-Oxazolidinone

98%

Synonym(s):

1,3-Oxazolidin-2-one, 2-Oxazolidone, 2-Oxo-1,3-oxazolidine, 2-Oxotetrahydro-1,3-oxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NO2
CAS Number:
Molecular Weight:
87.08
Beilstein:
106251
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

220 °C/48 mmHg (lit.)

mp

83-87 °C (lit.)

SMILES string

O=C1NCCO1

InChI

1S/C3H5NO2/c5-3-4-1-2-6-3/h1-2H2,(H,4,5)

InChI key

IZXIZTKNFFYFOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Oxazolidinone can be employed as a precursor for the preparation of acryloyloxazolidin-2-ones , trans-3-cinnamoyloxazolidin-2-one, 3-bromo-2-oxazolidinone , 4-methoxy-2-oxazolidinone , coordination compound, tetrakis[μ-(2-oxazolidinonato-κN3:κO2)]tetra-, (4Cu-Cu).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Diels-Alder Reaction of α,β -Unsaturated Oxazolidin-2-one Derivatives Catalyzed by a Chiral Fe (III)-Bipyridine Diol Complex
Li Mao, et al.
Organic Letters, 20(4), 995-998 (2018)
Cysteine-Aminoethylation-Assisted Chemical Ubiquitination of Recombinant Histones
Chu G-C, et al.
Journal of the American Chemical Society, 141(8), 3654-3663 (2019)
Stereocontrolled Synthesis of trans/cis-2, 3-Disubstituted Cyclopropane-1, 1-diesters and Applications in the Syntheses of Furanolignans
Shen Y, et al.
The Journal of Organic Chemistry, 83(20), 12549-12558 (2018)
Mathieu Esgulian et al.
Organic & biomolecular chemistry, 15(6), 1453-1462 (2017-01-21)
A number of cyclic derivatives of 3-amino-2,4-dihydroxybutanoic acid are known in the literature but they are often prepared from other cyclic precursors. This study showed that the title compound too may serve as a convenient substrate for cyclization reactions. Using
Nada A Al-Hasawi et al.
Anticancer research, 40(9), 5125-5140 (2020-09-04)
Neuroblastoma (NB), the most common extracranial malignant childhood tumor accounts for about 15% of cancer-related deaths in children. Despite the intensive treatment of patients with high-risk scarification of NB, clinical outcomes indicate tumor recurrence greater than 50% and late severe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service