Skip to Content
Merck
All Photos(2)

Key Documents

M72803

Sigma-Aldrich

2-Methylpiperidine

98%

Synonym(s):

α-Pipecoline, 2-Pipecoline, NSC 31047, NSC 462

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
Beilstein:
79804
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

118-119 °C/753 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CC1CCCCN1

InChI

1S/C6H13N/c1-6-4-2-3-5-7-6/h6-7H,2-5H2,1H3

InChI key

NNWUEBIEOFQMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation

Reactant for synthesis of:
  • Azepan-4-ones via two step [5+2] annulation
  • 2-Aminobenzoxazoles
  • Unsymmetrically substituted ureas
  • Corticotropin-releasing factor receptor type 1 antagonists
  • Gefitinib analogues with anti-tumor activity

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaoqing Wu et al.
Bioorganic & medicinal chemistry, 18(11), 3812-3822 (2010-05-15)
There is an urgent need to design and develop new and more potent EGFR inhibitors with improved anti-tumor activity. Here we describe the design and synthesis of two series of 4-benzothienyl amino quinazolines as new analogues of the EGFR inhibitor
Li Cui et al.
Chemical communications (Cambridge, England), 46(19), 3351-3353 (2010-05-06)
A surprisingly efficient synthesis of azepan-4-ones via a two-step [5 + 2] annulation is developed. This reaction involves a key gold catalysis and shows generally high regioselectivities and good to excellent diastereoselectivities.
Christopher L Cioffi et al.
The Journal of organic chemistry, 75(22), 7942-7945 (2010-10-27)
The synthesis of 2-aminobenzoxazoles can be readily achieved by two versatile, one-pot procedures utilizing commercially available tetramethyl orthocarbonate or 1,1-dichlorodiphenoxymethane, an amine, and an optionally substituted 2-aminophenol. The reactions were conducted under mild conditions and provided 2-aminobenzoxazoles in modest to
Dmitry Zuev et al.
Bioorganic & medicinal chemistry letters, 20(12), 3669-3674 (2010-05-18)
A novel series of [6-chloro-2-trifluoromethyl-7-aryl-7H-imidazo[1,2-a]imidazol-3-ylmethyl]-dialkylamines was discovered as potent CRF(1)R antagonists. The optimization of binding affinity in the series by the parallel reaction approach is discussed herein.
C-2 arylation of piperidines through directed transition-metal-catalyzed sp3 C-H activation.
Hana Prokopcová et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(44), 13063-13067 (2010-10-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service