Skip to Content
Merck
All Photos(3)

Key Documents

M6204

Sigma-Aldrich

2-Thiazoline-2-thiol

98%

Synonym(s):

2-Mercapto-2-thiazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NS2
CAS Number:
Molecular Weight:
119.21
Beilstein:
106332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

100-105 °C (lit.)

SMILES string

S=C1NCCS1

InChI

1S/C3H5NS2/c5-3-4-1-2-6-3/h1-2H2,(H,4,5)

InChI key

WGJCBBASTRWVJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tool for highly selective chiral syntheses of penam- and carbapenam-type β-lactam antibiotics.[1][2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stud. Org. Chem., 28, 57-57 (1987)
Yahia N Mabkhot et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-01)
A series of new thiazoline derivatives were synthesized. Structure analyses were accomplished employing 1H-NMR, 13C-NMR, X-ray and MS techniques. The in vitro antitumor activities were assessed against human hepatocellular carcinoma (HepG-2) and colorectal carcinoma (HCT-116) cell lines. The results revealed
Cestmír Konák et al.
Langmuir : the ACS journal of surfaces and colloids, 24(14), 7092-7098 (2008-06-12)
Vesicles bearing either cationic (amino) groups or zwitterionic (amino acid) groups on the surface were coated with a reactive multivalent hydrophilic N-(2-hydroxypropyl)methacrylamide polymer (PHPMA) and its positively charged analogue (3 mol % quaternary ammonium groups), both having reactive thiazolidine-2-thione (TT)
Taro Hase et al.
Urology, 60(3), 542-547 (2002-09-28)
To investigate the expression of peroxisome proliferator activator-receptor (PPAR)-alpha, beta, and gamma in human testicular cancer (TC) and normal testicular (NT) tissues, as well as the effects of the PPAR-gamma ligand. Recent studies have demonstrated that PPAR-gamma is expressed in
R B Greenwald et al.
Bioconjugate chemistry, 7(6), 638-641 (1996-11-01)
A novel PEG linker that employs a thiazolidine-2-thione group has been synthesized. Kinetic studies done on this compound demonstrate a relatively long half-life compared to those of traditional succinimidyl linkers. This new PEG derivative reacts with proteins under mild conditions

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service