Skip to Content
Merck
All Photos(2)

Documents

M28804

Sigma-Aldrich

2-(2-Methylaminoethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.518 (lit.)

bp

113-114 °C/30 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

CNCCc1ccccn1

InChI

1S/C8H12N2/c1-9-7-5-8-4-2-3-6-10-8/h2-4,6,9H,5,7H2,1H3

InChI key

UUQMNUMQCIQDMZ-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Brahim Tighilet et al.
Frontiers in neurology, 9, 431-431 (2018-06-27)
This study is a pharmacokinetic (PK) and pharmacodynamics (PD) approach using betahistine doses levels in unilateral vestibular neurectomized cats (UVN) comparable to those used in humans for treating patients with Menière's disease. The aim is to investigate for the first
Mattis Bertlich et al.
Life sciences, 187, 17-21 (2017-08-19)
Betahistine is a histamine analogue that is used for the treatment of Menière's disease. Animal studies showed that it increases local blood flow in the stria vascularis. In terms of its mode of action, recent studies have prompted discussion of
Adriana Pontin Garcia et al.
Brazilian journal of otorhinolaryngology, 79(3), 366-374 (2013-06-08)
Virtual reality technology can provide a wide range of sensory stimuli to generate conflicts of varying degrees of complexity in a safe environment. To verify the effect of a virtual realitybased balance rehabilitation program for patients with Menière's disease. This
Hiroshi Ueno et al.
The Libyan journal of medicine, 15(1), 1808361-1808361 (2020-08-19)
The administration of therapeutic agents is difficult in many patients, such as patients with post-operative delirium or dementia or patients with schizophrenia, who are upset in an emergency room. Therefore, the development of a new method for administering therapeutic agents
Maurício Malavasi Ganança et al.
Brazilian journal of otorhinolaryngology, 77(4), 499-503 (2011-08-24)
Betahistine is a medicine used to treat vestibular disorders that has also been used to treat tinnitus. To assess the effects of betahistine on tinnitus in patients with vestibular disorders. Retrospective data were collected from patient records for individuals presenting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service