M14935
5-Methoxy-3-indoleacetic acid
98%
Synonym(s):
2-(5-Methoxy-3-indolyl)acetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H11NO3
CAS Number:
Molecular Weight:
205.21
Beilstein:
187161
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
145-148 °C (dec.) (lit.)
SMILES string
COc1ccc2[nH]cc(CC(O)=O)c2c1
InChI
1S/C11H11NO3/c1-15-8-2-3-10-9(5-8)7(6-12-10)4-11(13)14/h2-3,5-6,12H,4H2,1H3,(H,13,14)
InChI key
COCNDHOPIHDTHK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Reactant for preparation of aryloxybenzothiazoles as inhibitors of NO production
- Reactant for preparation of transcription inhibitors of novel Gli1 as antitumor agents
- Reactant for preparation of indole derivatives as NR2B/NMDA receptor antagonists
- Reactant for preparation of indolyl esters and amides related to indomethacin as selective COX-2 inhibitors
- Reactant for preparation of quinoline salicylic acid series of P-selectin antagonists
- Reactant for preparation of prostaglandin D2 receptor antagonists
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
E S Rom-Bugoslavskaja et al.
Acta physiologica Hungarica, 70(4), 397-401 (1987-01-01)
In intact Wistar pubertal male rats held on LD 8:16 (winter) and 16:8 (summer) pineal melatonin (M) production and other pineal indoles content serotonin (S), 5-methoxytriptamine (5-MT), N-acetylserotonin (N-aS), 5-hydroxyindoleacetic acid (5-HIAA) and 5-methoxyindoleacetic acid (5-MIAA) were investigated in basic
H Meissl et al.
Journal of pineal research, 17(2), 69-78 (1994-09-01)
The effect of benzodiazepines (BZP) on melatonin release was investigated in the pineal gland of the rainbow trout, Oncorhynchus mykiss, maintained under in vitro perifusion culture conditions. Melatonin and the methoxyindoles 5-methoxytryptophol (5-MTOL), 5-methoxyindoleacetic acid (5-MIAA), and 5-methoxytryptamine (5-MT) were
J Yáñez et al.
Neurochemistry international, 27(2), 195-200 (1995-08-01)
Synthesis and release of the pineal hormone melatonin is in all vertebrates primarily regulated by the light/dark cycle. In pineal organs of teleost fish, like in other non-mammalian vertebrates, melatonin formation is regulated by a direct photoreception of the pineal
John M Wilson et al.
Forensic science international, 148(1), 31-36 (2004-12-21)
Foxy is the colloquial name for the hallucinogen 5-ethoxy-diisopropyltryptamine (5-MeO-DIPT). A non-fatality involving a 23-year-old Caucasian man who ingested a capsule containing 5-MeO-DIPT is described. He presented to the Emergency Department, not with visual nor auditory hallucinations but with sensory
M S Grace et al.
Neuroscience, 62(2), 615-623 (1994-09-01)
Melatonin modulates a variety of rhythmic processes in vertebrates, and is synthesized in both the retina and pineal gland. We have shown previously that retinal melatonin is deacetylated generating 5-methoxytryptamine, which is then deaminated by monoamine oxidase, producing 5-methoxyindoleacetic acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service