Skip to Content
Merck
All Photos(3)

Key Documents

K6625

Sigma-Aldrich

2-Ketohexanoic acid sodium salt

97.0-103.0%

Synonym(s):

α-Ketocaproic acid sodium salt, 2-Ketocaproic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9O3Na
CAS Number:
Molecular Weight:
152.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97.0-103.0%

form

powder

mp

240 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Na].CCCCC(=O)C(O)=O

InChI

1S/C6H10O3.Na/c1-2-3-4-5(7)6(8)9;/h2-4H2,1H3,(H,8,9);/q;+1/p-1

InChI key

WDCARDDLMCHULC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Salehi et al.
Cellular signalling, 10(9), 645-651 (1998-10-30)
We examined the relation between the islet NO system and islet hormone secretion induced by either the non-glucose nutrient alpha-ketoisocaproic acid (KIC) or, in some experiments, glucose. KIC dose dependently stimulated insulin but inhibited glucagon secretion. In a medium devoid
S Vijayaraghavan et al.
Biology of reproduction, 40(4), 744-751 (1989-04-01)
Immature caput epididymal sperm accumulate calcium from exogenous sources at a rate 2- to 4-fold greater than mature caudal sperm. Calcium accumulation by these cells, however, is maximal in the presence of lactate as external substrate. This stimulation of calcium
T Sekimoto et al.
Journal of biochemistry, 116(1), 176-182 (1994-07-01)
A tetrapeptide sequence, Gly-Gly-(Gly/Ala)-Lys, containing a catalytically important lysyl residue, is highly conserved in NAD(P)+-dependent amino acid dehydrogenases. To elucidate functional roles of the glycyl residues in this conserved sequence Gly-77, Gly-78, and Gly-79 of the recombinant leucine dehydrogenase from
G P Butrimovitz et al.
Clinical chemistry, 29(8), 1518-1521 (1983-08-01)
We have developed an automated enzymatic assay for LDH-C4, a lactate dehydrogenase (EC 1.1.1.27) isoenzyme found exclusively in spermatozoa, as a marker of human spermatogenesis. LDH-C4 activity is assayed bichromatically in the Abbott ABA-100 spectrophotometer, with 2-oxohexanoate as substrate. As
G L Loy et al.
Analytical biochemistry, 185(1), 1-9 (1990-02-15)
Tracer methods using both carbon-13 and -14 have been utilized for determination of ovine fetal amino acid disposal and the results compared in seven animals. We infused [1-13C]leucine simultaneously with [1-14C]leucine into the fetal circulation of pregnant sheep chronically catheterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service