I17907
4-Piperidinecarboxamide
97%
Synonym(s):
Hexahydroisonicotinamide, Isonipecotamide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H12N2O
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
145-148 °C (lit.)
SMILES string
NC(=O)C1CCNCC1
InChI
1S/C6H12N2O/c7-6(9)5-1-3-8-4-2-5/h5,8H,1-4H2,(H2,7,9)
InChI key
DPBWFNDFMCCGGJ-UHFFFAOYSA-N
Application
Reactant for synthesis of:
Reactant for identification of small molecular inhibitors of importin-β mediated nublear import
- Survival motor neuron (SMN) protein modulators
- Diaminotriazine hNav1.7 inhibitors
- Heteroalicyclic carboxamidines as inhibitors of inducible nitric oxide synthase
- Orally available naphthyridine protein kinase D inhibitors
- Phosphodiesterase 5 inhibitors
Reactant for identification of small molecular inhibitors of importin-β mediated nublear import
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service