Skip to Content
Merck
All Photos(1)

Key Documents

H49901

Sigma-Aldrich

3-Hydroxyphenylacetic acid

≥99%

Synonym(s):

(3-Hydroxyphenyl)acetic acid, (m-Hydroxyphenyl)acetic acid, 2-(3-Hydroxyphenyl)acetic acid, 2-(3′-Hydroxyphenyl)acetic acid, 3-Hydroxybenzeneacetic acid, 3′-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2086506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

mp

129-133 °C (lit.)

SMILES string

OC(=O)Cc1cccc(O)c1

InChI

1S/C8H8O3/c9-7-3-1-2-6(4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)

InChI key

FVMDYYGIDFPZAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doris M Jacobs et al.
Journal of agricultural and food chemistry, 60(12), 3078-3085 (2012-03-01)
Red wine and grape polyphenols are considered to promote cardiovascular health and are involved in multiple biological functions. Their overall impact on the human metabolome is not known. Therefore, exogenous and endogenous metabolic effects were determined in fasting plasma and
P S McQuade et al.
Progress in neuro-psychopharmacology & biological psychiatry, 7(4-6), 755-759 (1983-01-01)
The administration of deuterated beta-phenylethylamine to mice causes increased concentrations of deuterated para-hydroxyphenylacetic acid and meta-hydroxyphenylacetic acid within the caudate nuclei two hours after injection. Deuterated m-HPAA concentrations remain elevated at 4 hours. This suggests that high concentrations of PE
Xin Fang et al.
Frontiers in microbiology, 9, 2559-2559 (2018-11-15)
Dysbiosis of the gut microbiome, including elevated abundance of putative leading bacterial triggers such as E. coli in inflammatory bowel disease (IBD) patients, is of great interest. To date, most E. coli studies in IBD patients are focused on clinical
Matthew Snelson et al.
Science advances, 7(14) (2021-04-02)
Intake of processed foods has increased markedly over the past decades, coinciding with increased microvascular diseases such as chronic kidney disease (CKD) and diabetes. Here, we show in rodent models that long-term consumption of a processed diet drives intestinal barrier
Hanne Winning et al.
The Analyst, 134(11), 2344-2351 (2009-10-20)
The metabolome following intake of onion by-products is evaluated. Thirty-two rats were fed a diet containing an onion by-product or one of the two derived onion by-product fractions: an ethanol extract and the residue. A 24 hour urine sample was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service