All Photos(1)
About This Item
Linear Formula:
CH3(CH2)14CH=CH2
CAS Number:
Molecular Weight:
238.45
Beilstein:
1758486
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.441 (lit.)
bp
157-159 °C/11 mmHg (lit.)
mp
10-11 °C (lit.)
density
0.785 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCC=C
InChI
1S/C17H34/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h3H,1,4-17H2,2H3
InChI key
ADOBXTDBFNCOBN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Changhao Bi et al.
Microbial cell factories, 12, 107-107 (2013-11-14)
The chemoautotrophic bacterium Ralstonia eutropha can utilize H2/CO2 for growth under aerobic conditions. While this microbial host has great potential to be engineered to produce desired compounds (beyond polyhydroxybutyrate) directly from CO2, little work has been done to develop genetic
Ian Sofian Yunus et al.
Metabolic engineering, 49, 201-211 (2018-08-26)
Liquid fuels sourced from fossil sources are the dominant energy form for mobile transport today. The consumption of fossil fuels is still increasing, resulting in a continued search for more sustainable methods to renew our supply of liquid fuel. Photosynthetic
Jaimie Gibbons et al.
AMB Express, 8(1), 167-167 (2018-10-15)
Cyanobacteria photosynthetically produce long-chain hydrocarbons, which are considered as infrastructure-compatible biofuels. However, native cyanobacteria do not produce these hydrocarbons at sufficient rates or yields to warrant commercial deployment. This research sought to identify specific genes required for photosynthetic production of
Zihao Zhang et al.
ChemSusChem, 13(18), 4922-4928 (2020-07-17)
Catalytic deoxygenation of even-numbered fatty acids into odd-chain linear α-olefins (LAOs) has emerged as a complementary strategy to oligomerization of ethylene, which only affords even-chain LAOs. Although enzymes and homogeneous catalysts have shown promising potential for this application, industrial production
Roberto Barone et al.
Marine drugs, 18(3) (2020-03-22)
Algae have multiple similarities with fungi, with both belonging to the Thallophyte, a polyphyletic group of non-mobile organisms grouped together on the basis of similar characteristics, but not sharing a common ancestor. The main difference between algae and fungi is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service