Skip to Content
Merck
All Photos(2)

Key Documents

GF95821195

Carbon

rod, 50mm, diameter 50mm, graphite, 99.95%

Synonym(s):

Activated charcoal, C 007975, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.95%

form

rod

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 958-211-95

resistivity

1375 μΩ-cm, 20°C (graphite)

size

50 × 50 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elson C Santos et al.
Journal of nanoscience and nanotechnology, 14(6), 4590-4594 (2014-04-18)
The system proposed consists of a flagellum relaxing around a static carbon nanowire to mimics behavior of a natural flagellum moving with damped harmonic motion along a wire under van der Waals and electrostatic forces. This flagellum is composed of
Chi-lai Ho et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 749-752 (2014-03-29)
We prospectively compared (11)C-acetate with (18)F-FDG in a PET/CT evaluation of multiple myeloma (MM), specifically on diagnostic accuracy, identification of high-risk patients, and monitoring of treatment response. Dual-tracer PET/CT was performed on 35 pathologically and clinically confirmed and untreated patients
Anna Iliadi et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 425-433 (2014-04-08)
The aim of this study was to assess the effect of intraoral aging on the setting status of a resin composite and a glass ionomer adhesive, relative to control specimens stored in water. Metallic brackets were bonded with resin composite
Cancer treatment: Sharp shooters.
Vivien Marx
Nature, 508(7494), 133-138 (2014-04-04)
B Mukherjee et al.
Journal of environmental biology, 35(3), 577-588 (2014-05-13)
Modelling system dynamics in a hyper-eutrophic lake is quite complex especially with a constant influx of detergents and sewage material which continually changes the state variables and interferes with the assessment of the chemical rhythm occurring in polluted conditions as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service