GF85108293
Aluminum-silicon alloy, Al88Si12
200mm rod, 15mm diameter, extruded
Synonym(s):
AL137920
About This Item
Recommended Products
form
rod
manufacturer/tradename
Goodfellow 851-082-93
L × diam.
200 mm × 15 mm
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | GF45484836 | GF92936919 | GF28602503 |
---|---|---|---|
Goodfellow 851-082-93 | Goodfellow 454-848-36 | Goodfellow 929-369-19 | Goodfellow 286-025-03 |
200 mm × 15 mm | 500 mm × 15 mm | - | - |
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service