GF57863931
Rhenium
wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Synonym(s):
Rhenium, RE005120
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99.97%
form
wire
manufacturer/tradename
Goodfellow 578-639-31
L × diam.
0.1 m × 0.25 mm
bp
5596 °C (lit.)
5627 °C (lit.)
mp
3180 °C (lit.)
density
21.02 g/cm3 (lit.)
SMILES string
[ReH]
InChI
1S/Re
InChI key
WUAPFZMCVAUBPE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rheumatology (Oxford, England), 46(10), 1531-1537 (2007-06-29)
Hypertrophy and inflammation of the synovium with various underlying pathologies - such as rheumatoid arthritis, osteoarthritis, haemophilia and spondyloarthropathy - can be treated successfully by radiosynoviorthesis (RSO). For medium-sized joints (shoulder, elbow, wrist, hip and ankle), the radionuclide of choice
Mini reviews in medicinal chemistry, 4(1), 61-68 (2004-02-03)
The favourable nuclear properties of copper-67 and rhenium-188 for therapeutic application are described, together with methods for the chemical synthesis of a number of derivatives. Survival from invasive bladder cancer has changed little over the past 20 years. The intravesicular
Radiolabeling of monoclonal antibodies and fragments with technetium and rhenium.
Bioconjugate chemistry, 3(2), 91-99 (1992-03-01)
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 54(3), 399-406 (2001-02-24)
188Re is one of the radioisotopes expected to emerge as useful for therapy. Development of new radiopharmaceuticals based on 188Re depends on the radiolabeling methods used, which would give stable complexes having predefined radiochemical properties and in vitro and in
Chemical communications (Cambridge, England), (5)(5), 493-512 (2009-03-14)
Radiolabeled biomolecules can be used to visualize a variety of diseases through interaction with specific cell receptors. A key step is the introduction of a molecular entity that allows facile labeling with the medically useful radionuclide (99m)Tc without significant alteration
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service