GF49993048
Molybdenum
microfoil, disks, 25mm, thinness 1.0μm, specific density 1022μg/cm2, permanent mylar 3.5μm support, 99.9%
Synonym(s):
Molybdenum, MO004850
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99.9%
form
foil
manufacturer/tradename
Goodfellow 499-930-48
resistivity
5.0 μΩ-cm, 20°C
diam. × thickness
25 mm × 1.0 μm
bp
4612 °C (lit.)
mp
2617 °C (lit.)
density
10.3 g/mL at 25 °C (lit.)
SMILES string
[Mo]
InChI
1S/Mo
InChI key
ZOKXTWBITQBERF-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Metal ions in life sciences, 13, 415-450 (2014-01-29)
Molybdenum is an essential trace element and crucial for the survival of animals. Four mammalian Mo-dependent enzymes are known, all of them harboring a pterin-based molybdenum cofactor (Moco) in their active site. In these enzymes, molybdenum catalyzes oxygen transfer reactions
The Science of the total environment, 435-436, 96-106 (2012-08-01)
In a regulatory context, bioaccumulation or bioconcentration factors are used for considering secondary poisoning potential and assessing risks to human health via the food chain. In this paper, literature data on the bioaccumulation of molybdenum in the aquatic organisms are
Advances in experimental medicine and biology, 675, 49-70 (2010-06-10)
The vast majority of the purple nonsulfur photosynthetic bacteria are diazotrophs, but the details of the complex regulation of the nitrogen fixation process are well understood only for a few species. Here we review what is known of the well-studied
Journal of cellular biochemistry, 112(10), 2721-2728 (2011-06-17)
When intracelluar pathogens enter the host macrophages where in addition to oxidative and antibiotic mechanisms of antimicrobial activity, nutrients are deprived. Human pathogen Mycobacterium tuberculosis is one of macrophage parasitisms, which can replicate and persist for decades in dormancy state
Journal of nanoscience and nanotechnology, 14(6), 4462-4468 (2014-04-18)
A mild one-pot hydrothermal route has been successfully designed to controllably prepare orthorhombic alpha-MoO3 nanobelts and monoclinic MoO2 microaxletrees respectively by adjusting the dosage of (NH4)6M07O24 x 4H2O (AHM). The products are characterized by X-ray diffraction (XRD), field-emission scanning electron
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service