GF41758768
Molybdenum
foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Synonym(s):
Molybdenum, MO000170
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23
Assay
99.9%
form
foil
manufacturer/tradename
Goodfellow 417-587-68
resistivity
5.0 μΩ-cm, 20°C
bp
4612 °C (lit.)
mp
2617 °C (lit.)
density
10.3 g/mL at 25 °C (lit.)
SMILES string
[Mo]
InChI
1S/Mo
InChI key
ZOKXTWBITQBERF-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P Kielczynski et al.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control, 46(1), 233-238 (2008-02-02)
In this paper the theoretical analysis and the results of testing of a piezoelectric cantilever for the investigation of material surfaces are presented. The cantilever consists of a thin piezoelectric plate bonded with a thin metal (e.g., molybdenum) foil. The
Charles F Baker et al.
Dalton transactions (Cambridge, England : 2003), (9)(9), 1298-1303 (2004-07-15)
New molybdenum(VI) nitride oxides were synthesised by the reaction of strontium nitride and calcium nitride with molybdenum foil at high temperature in sealed stainless steel crucibles. The reactions yielded single crystalline products determined by X-ray diffraction to form complex structures
Dattatray J Late et al.
Nanotechnology, 19(26), 265605-265605 (2008-07-02)
Nanocrystalline lanthanum hexaboride (LaB(6)) films have been deposited on molybdenum foil by the pulsed laser deposition (PLD) technique. The as-deposited films were characterized by x-ray diffraction (XRD), scanning electron microscopy (SEM) and atomic force microscopy (AFM). The XRD pattern shows
J G Watkins et al.
The Review of scientific instruments, 79(10), 10F125-10F125 (2008-12-03)
Two modular arrays of Langmuir probes designed to handle a heat flux of up to 25 MW/m(2) for 10 s exposures have been installed in the lower divertor target plates of the DIII-D tokamak. The 20 pyrolytic graphite probe tips
Michael N Evans
Rapid communications in mass spectrometry : RCM, 22(14), 2211-2219 (2008-06-11)
A reactor for converting cellulose into carbon monoxide for subsequent oxygen isotopic analysis via continuous flow isotope ratio mass spectrometry is described. The system employs an induction heater to produce temperatures >or=1500 degrees C within a molybdenum foil crucible positioned
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service