Skip to Content
Merck
All Photos(2)

Key Documents

GF39171517

Terbium

foil, 50x50mm, thickness 0.1mm, as rolled, 99%

Synonym(s):

Terbium, TB000210

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Tb
CAS Number:
Molecular Weight:
158.93
MDL number:
UNSPSC Code:
12141614
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 391-715-17

resistivity

116 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.1 mm

bp

3230 °C (lit.)

mp

1356 °C (lit.)

density

8.234 g/mL at 25 °C (lit.)

SMILES string

[Tb]

InChI

1S/Tb

InChI key

GZCRRIHWUXGPOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi Shi et al.
Cancer research, 75(1), 147-158 (2014-11-02)
Cables1 is a candidate tumor suppressor that negatively regulates cell growth by inhibiting cyclin-dependent kinases. Cables1 expression is lost frequently in human cancer but little is known about its regulation. Here, we report that Cables1 levels are controlled by a
Dinari A Harris et al.
Current protocols in nucleic acid chemistry, Chapter 6, Unit 6-Unit 6 (2008-04-23)
The function of an RNA molecule is determined by its overall secondary and tertiary structure. The tertiary structure is facilitated and stabilized by the interaction with metal ions. The current chapter offers a detailed protocol on the use of the
Lei Rao et al.
International journal of food microbiology, 205, 73-80 (2015-04-19)
The objective of this study was to investigate the inactivation of the Bacillus subtilis spores by high pressure CO2 combined with high temperature (HPCD+HT) and to analyze the clumping effect of the spores on their HPCD+HT resistance. The spores of
Margaret L Warner et al.
Molecular pharmacology, 87(3), 410-420 (2014-12-17)
Secreted R-spondin proteins (RSPOs1-4) function as adult stem cell growth factors by potentiating Wnt signaling. Simultaneous binding of distinct regions of the RSPO Fu1-Fu2 domain module to the extracellular domains (ECDs) of the LGR4 G protein-coupled receptor and the ZNRF3
Kamal P Mani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 148, 412-419 (2015-04-29)
Terbium molybdate nanophosphors were synthesized through a facile sol-gel route. The structure of the phosphors was characterized by X-ray diffraction, Raman spectra and Fourier transform infrared spectroscopy analysis. The X-ray diffraction studies revealed that the structure of the nanophosphor gradually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service