GF23496756
Tantalum
foil, 50mm disks, thickness 0.03mm, as rolled, 99.9%
Synonym(s):
Tantalum, TA000348, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.90%
form
foil
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 234-967-56
resistivity
13.5 μΩ-cm, 20°C
diam. × thickness
50 mm × 0.03 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tantalum foil used in closing antro-oral fistulas.
E J McCLUNG et al.
United States Armed Forces medical journal, 2(8), 1183-1186 (1951-08-01)
Isolation of the brachial segments of the spinal cord of the chick embryo by means of tantalum foil blocks.
V HAMBURGER
The Journal of experimental zoology, 103(1), 113-142 (1946-10-01)
H H Pieper et al.
The Review of scientific instruments, 83(5), 055110-055110 (2012-06-07)
We present a versatile system for the preparation of oxide crystal surfaces in the ultra-high vacuum (UHV) at temperatures up to 1300 K. Thermal treatment is accomplished by direct current heating of a tantalum foil in contact with the oxide
Arthroplasty of the temporomandibular joint in children with interposition of tantalum foil.
G W N EGGERS
The Journal of bone and joint surgery. American volume, 28, 603-606 (1946-07-01)
Xinjian Feng et al.
Nano letters, 10(3), 948-952 (2010-02-12)
Tantalum nitride (Ta3N5) has a band gap of approximately 2.07 eV, suitable for collecting more than 45% of the incident solar spectrum energy. We describe a simple method for scale fabrication of highly oriented Ta3N5 nanotube array films, by anodization
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service