Skip to Content
Merck
All Photos(2)

Key Documents

GF14894163

Rhenium

wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%

Synonym(s):

Rhenium, RE005105

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12141737
PubChem Substance ID:
NACRES:
NA.23

Assay

99.97%

form

wire

manufacturer/tradename

Goodfellow 148-941-63

L × diam.

0.5 m × 0.076 mm

bp

5596 °C (lit.)
5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malcolm Frier
Mini reviews in medicinal chemistry, 4(1), 61-68 (2004-02-03)
The favourable nuclear properties of copper-67 and rhenium-188 for therapeutic application are described, together with methods for the chemical synthesis of a number of derivatives. Survival from invasive bladder cancer has changed little over the past 20 years. The intravesicular
R Klett et al.
Rheumatology (Oxford, England), 46(10), 1531-1537 (2007-06-29)
Hypertrophy and inflammation of the synovium with various underlying pathologies - such as rheumatoid arthritis, osteoarthritis, haemophilia and spondyloarthropathy - can be treated successfully by radiosynoviorthesis (RSO). For medium-sized joints (shoulder, elbow, wrist, hip and ankle), the radionuclide of choice
Radiolabeling of monoclonal antibodies and fragments with technetium and rhenium.
G L Griffiths et al.
Bioconjugate chemistry, 3(2), 91-99 (1992-03-01)
Peter Wolohan et al.
Steroids, 72(3), 247-260 (2007-02-07)
OPLS all atom force field parameters were developed in order to model a diverse set of novel rhenium based estrogen receptor ligands whose relative binding affinities (RBA) to the estrogen receptor alpha isoform (ERalpha) with respect to 17beta-estradiol were available.
N Iznaga-Escobar
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 54(3), 399-406 (2001-02-24)
188Re is one of the radioisotopes expected to emerge as useful for therapy. Development of new radiopharmaceuticals based on 188Re depends on the radiolabeling methods used, which would give stable complexes having predefined radiochemical properties and in vitro and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service