Skip to Content
Merck
All Photos(2)

Documents

GF04790033

Nickel

foil, 4mm disks, thickness 0.25mm, as rolled, 100%

Synonym(s):

Nickel, NI000565

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

100%

form

foil

manufacturer/tradename

Goodfellow 047-900-33

resistivity

6.97 μΩ-cm, 20°C

diam. × thickness

4 mm × 0.25 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G E van Dorssen et al.
Journal of X-ray science and technology, 3(2), 109-117 (1992-01-01)
The structure of Ni-C multilayer and single nickel layer samples has been analyzed before and after annealing, using two techniques: fluorescence EXAFS (F1EXAFS) at the Ni-K. edge and CuKα reflection. Annealing at a temperature of 450°C resulted in a change
F E Cecil et al.
The Review of scientific instruments, 81(10), 10D326-10D326 (2010-11-09)
We have examined the observed currents in the front foils of the JET Faraday cup lost alpha particle diagnostic KA-2. In particular, we have sought to understand the currents during Ohmic plasmas for which the ion flux at the detectors
W K Winnett et al.
Talanta, 41(10), 1627-1630 (1994-10-01)
A modification of the sample introduction system for combustion total organic carbon analyzers is presented where sample is sprayed into the instrument. Thermally created sprays and nebulizer sprays are discussed. It is shown that spray introduction gives more symmetric signals
L A van Loon et al.
Journal of oral pathology, 17(3), 129-137 (1988-03-01)
An in vivo comparison was made between the contact allergic stomatitis-inducing capacity of nickel, nickel-containing dental alloys and a non-corrosive precious metal. Fifteen patients with a positive allergic skin reaction to nickel were divided into 3 groups (A, B and
Gareth E Haslam et al.
Physical chemistry chemical physics : PCCP, 13(28), 12968-12974 (2011-06-23)
Metallic nickel is a powerful electrocatalyst in alkaline solution and is able to be used in the alkaline fuel cell. However, in acidic solution, electrocatalysis is impossible because the metal is subject to rapid corrosion at low pH for all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service