Skip to Content
Merck
All Photos(1)

Key Documents

G5908

Sigma-Aldrich

(±)-Geosmin solution

2 mg/mL in methanol, ≥97%

Synonym(s):

DL-Geosmin, rel-(4R,4aR,8aS)-Octahydro-4,8a-dimethyl-4a(2H)-naphthalenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (calc. on dry substance, GC)
≥97%

concentration

2 mg/mL in methanol

storage temp.

−20°C

SMILES string

OC21C(CCCC2C)(CCCC1)C

InChI

1S/C12H22O/c1-10-6-5-8-11(2)7-3-4-9-12(10,11)13/h10,13H,3-9H2,1-2H3

InChI key

JLPUXFOGCDVKGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Geosmin solution is commonly used in the odorant removal/degradation studies in the water and food chemistry.[1][2][3]

Other Notes

The (−)-enantiomer is the principal component of beet aroma.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of 2-MIB and geosmin using UV/persulfate: contributions of hydroxyl and sulfate radicals.
Xie, Pengchao et al.
Water Research, 69(6), 223-233 (2015)
Buttery, R.G., et al.
Journal of Agricultural and Food Chemistry, 24, 419 and 1246-419 and 1246 (1976)
Development of long-life-cycle tablet ceramic adsorbent for geosmin removal from water solution.
Chen, Rongzhi aet al.
Applied Surface Science, 257(6), 2091-2096 (2011)
Shuke Xiao et al.
eLife, 8 (2019-10-28)
Odorant binding proteins (Obps) are expressed at extremely high levels in the antennae of insects, and have long been believed essential for carrying hydrophobic odorants to odor receptors. Previously we found that when one functional type of olfactory sensillum in
Stable TiO 2-USY zeolite composite coatings for efficient adsorptive and photocatalytic elimination of geosmin from water.
Wee, Lik H et al.
Journal of Material Chemistry A, 3(5), 2258-2264 (2015)

Protocols

Separation of 2-Isopropyl-3-methoxypyrazine 20 ppt; 2-Isobutyl-3-methoxypyrazine; 2-Methylisoborneol; (±)-Geosmin 10 ppt

Questions

1–2 of 2 Questions  
  1. What can you tell me about the stereochemistry of Sigma-Aldrich's (±)-Geosmin products?

    1 answer
    1. The geosmin products listed by Sigma-Aldrich (CAS Registry number [16423-19-1]) are synthetic, and would be exactly a 50:50 mixture of the (+) and (-) isomers. Specifically, the natural (-) isomer is (4S,4aS,8aR)-4,8a-Dimethyl-1,2,3,4,5,6,7,8-octahydronaphthalen-4a-ol. Therefore, the unnatural (+) isomer (the mirror image) is (4R,4aR,8aS)-4,8a-Dimethyl-1,2,3,4,5,6,7,8-octahydronaphthalen-4a-ol.This information has not been confirmed by a specific rotation measurement or chiral analysis, but is known based on the method of preparation.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service