DWKWGW020VTML
Azlon DIY Safety Wash Bottles with Driplok vapor venting
low-density polyethylene bottle, capacity 1000 mL
Synonym(s):
WGW020VTML
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
low-density polyethylene bottle
packaging
pkg of 5 ea
manufacturer/tradename
Azlon WGW020VTML
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
Application
Ideal for solvent and water dispensing within the laboratory
Features and Benefits
Supplied with adhesive hazard labels to customise use
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service