DWKWGF532P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture
label, Methanol, low-density polyethylene bottle, capacity 250 mL
Synonym(s):
WGF532P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
material
low-density polyethylene bottle
feature
label Methanol
packaging
pkg of 5 ea
manufacturer/tradename
Azlon WGF532P
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for methannol dispensing within the laboratory
Features and Benefits
Slim design for comfortable handling and ease of use
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service