Key Documents
DWKW216933
Wheaton wide-mouth bottle
clear soda-lime glass bottle, capacity (4,000 mL), white polypropylene cap, (PTFE faced LDPE liner), case of 4 ea
Synonym(s):
W216933
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
(PTFE faced LDPE liner)
PTFE liner
clear soda-lime glass bottle
white polypropylene cap
feature
with cap
with cap
packaging
case of 4 ea
manufacturer/tradename
WHEATON® W216933
bottle diam. × H
(157 mm) (256 mm)
capacity
(4,000 mL)
fitting
89-400
General description
The Wheaton wide-mouth bottle is the largest size available in Type III glass bottles, specifically designed for general sample storage. Its wide-mouth opening facilitates easy filling and pouring, making it convenient for handling various types of samples. Constructed from durable glass, this bottle provides excellent chemical resistance, ensuring the integrity of the stored contents. Ideal for laboratories and industrial applications, it offers a reliable solution for managing bulk samples and liquids efficiently.
Features and Benefits
• Available in larger bottle sizes• Wide mouth / neck• Clear, Type III soda-lime glass• Available with caps attached or without caps• HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification• Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)