DWKBWP2000AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap
round amber polypropylene bottle, capacity 2000 mL
Synonym(s):
BWP2000AP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
round amber polypropylene bottle
wide-neck
packaging
pkg of 5 ea
manufacturer/tradename
Azlon BWP2000AP
capacity
2000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for a wide range of light sensitive liquid or powder applications
Features and Benefits
Autoclavable at 121°C and suitable for food contact applications
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service