DWKBWP1000PGRAD
Azlon Polypropylene Graduated Wide Neck Round Bottle with Write-On Panel
round translucent polypropylene bottle, screw cap, capacity 1000 mL
Synonym(s):
BWP1000PGRAD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
round translucent polypropylene bottle
screw cap
wide-neck
packaging
pkg of 5 ea
manufacturer/tradename
Azlon BWP1000PGRAD
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for a wide range of storage and preparation applications
Features and Benefits
Autoclavable at 121°C and suitable for food contact applications
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service