Skip to Content
Merck
All Photos(1)

Key Documents

D62403

Sigma-Aldrich

1,2-Dichloroethylene, mixture of cis and trans

98%

Synonym(s):

1,2-Dichloroethene, Acetylene dichloride, sym-Dichloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5.32 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

48-60 °C (lit.)

mp

−57 °C (lit.)

density

1.265 g/mL at 25 °C (lit.)

SMILES string

Cl\C=C\Cl

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1+

InChI key

KFUSEUYYWQURPO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloroethylene, also known as acetylene dichloride, is a chlorinated aliphatic hydrocarbon that is commonly used as an extraction solvent and as a chemical intermediate for other chlorinated solvents and compounds.

Application

1,2-Dichloroethylene, mixture of cis and trans can be used as a reactant to synthesize:
  • Chloro-enynes from terminal alkynes via Sonogashira coupling.
  • Trichloroethyl alkyl ketones by reacting with acyl chlorides via Friedel-Crafts reaction in the presence of aluminum chloride.
  • (Ethenediyl)bis[thiazole] via Pd-catalyzed Stille-coupling reaction with 2-(tributylstannyl)thiazole.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Idriss Curbet et al.
Chemical science, 11(19), 4934-4938 (2020-04-27)
The first synthesis of conjugated triynes by molybdenum-catalysed alkyne metathesis is reported. Strategic to the success of this approach is the utilization of sterically-hindered diynes that allowed for the site-selective alkyne metathesis to produce the desired conjugated triyne products. The
Dichloroethylene
James A and Mertens
Kirk-Othmer Encyclopedia of Chemical Technology (2000)
Unusual reaction of chloroacetyl chloride with 1, 2-dichloroethene. Synthesis and properties of 2-chlorovinyl dichloromethyl ketone
GV Bozhenkov, et al.
Russian Journal of Organic chemistry, 44, 1745-1751 (2008)
A quantum chemical study of unexpected reaction of alpha-chloroacyl chlorides with 1, 2-dichloroethylene in the presence of aluminum chloride
Shagun V, et al.
Computational & Theoretical Chemistry, 1073, 116-122 (2015)
Jisu Hong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(2), 649-656 (2018-11-06)
The effects of the molecular structure of thiazole-based polymers on the active layer morphologies and performances of electronic and photovoltaic devices were studied. Thus, thiazole-based conjugated polymers with a novel thiazole-vinylene-thiazole (TzVTz) structure were designed and synthesized. The TzVTz structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service