Skip to Content
Merck
All Photos(1)

Key Documents

D42003

Sigma-Aldrich

2,5-Dibromonitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3NO2
CAS Number:
Molecular Weight:
280.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

82-84 °C (lit.)

density

2.374 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(Br)ccc1Br

InChI

1S/C6H3Br2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H

InChI key

WRGKKASJBOREMB-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haluk Dinçalp et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 202, 196-206 (2018-05-23)
Five isoindigo-based donor-acceptor-donor (D-A-D) type small molecules have been synthesized in order to investigate their intramolecular charge transfer characteristics. UV-vis absorption of these dyes exhibits a wide absorption band ranging from 300 to 650 nm with two distinct bands, giving the
Derek E Williams et al.
Journal of the American Chemical Society, 140(24), 7611-7622 (2018-05-29)
Stimuli-responsive materials are vital for addressing emerging demands in the advanced technology sector as well as current industrial challenges. Here, we report for the first time that coordinative integration of photoresponsive building blocks possessing photochromic spiropyran and diarylethene moieties within
Ekaterina A Dolgopolova et al.
Journal of the American Chemical Society, 141(13), 5350-5358 (2019-03-07)
Electronic structure modulation of metal-organic frameworks (MOFs) through the connection of linker "wires" as a function of an external stimulus is reported for the first time. The established correlation between MOF electronic properties and photoisomerization kinetics as well as changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service