Skip to Content
Merck
All Photos(1)

Documents

D175706

Sigma-Aldrich

3,5-Dimethylphenol

98%

Synonym(s):

5-Hydroxy-m-xylene, sym.-m-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
774117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

(Powder or Crystals or Fibers or Chunk(s))

bp

222 °C (lit.)

mp

61-64 °C (lit.)
65-66 °C

SMILES string

Cc1cc(C)cc(O)c1

InChI

1S/C8H10O/c1-6-3-7(2)5-8(9)4-6/h3-5,9H,1-2H3

InChI key

TUAMRELNJMMDMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang-Jun Huo et al.
The Analyst, 136(9), 1892-1897 (2011-03-05)
A strategy for the determination of the presence of thiol-containing amino acids was successfully established by simply assembling copper chloride and xylenol orange (3,3'-bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein trisodium salt; XO) in a 1 : 1 molar ratio in quasi-physiological water solution (pH 6.0).
A M F Caldeira et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 65(5), 605-609 (2007-03-14)
Natural oxidation is an undesirable process suffered by gel dosimetry systems as it changes the values and spatial distribution of absorbed doses. Natural oxidation of Fricke xylenol gel (FXG) dosimeter was inferred from optical absorbance values measured for the first
E Södergren et al.
Journal of biochemical and biophysical methods, 37(3), 137-146 (1998-12-31)
The ferrous oxidation in xylenol orange version 2 (FOX2) assay coupled with triphenylphosphine has recently been employed for the measurement of total plasma hydroperoxides (ROOHs). In this study, we have evaluated sample handling and the effect of storage conditions on
R G Kelly et al.
Medical physics, 25(9), 1741-1750 (1998-10-17)
In recent years, magnetic-resonance imaging of gelatin doped with the Fricke solution has been applied to the direct measurement of three-dimensional (3D) radiation dose distributions. However, the 3D dose distribution can also be imaged more economically and efficiently using the
G Bieniek
Journal of chromatography. B, Biomedical applications, 682(1), 167-172 (1996-06-28)
An attempt was made to establish a method for the simultaneous determination of urinary concentrations of phenol, o-, p- and m-cresols, 1- and 2-naphthol and xylenol isomers by capillary gas chromatography. Urine samples were extracted after acid hydrolysis of glucuronides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service