Skip to Content
Merck
All Photos(3)

Key Documents

D16606

Sigma-Aldrich

6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate

99%

Synonym(s):

Acrinol, Ethacridine lactate salt, Ethodin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3O · C3H6O3 · H2O
CAS Number:
Molecular Weight:
361.39
Beilstein:
3642805
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

243-245 °C (lit.)

antibiotic activity spectrum

Gram-positive bacteria

SMILES string

O.CC(O)C(O)=O.CCOc1ccc2nc3cc(N)ccc3c(N)c2c1

InChI

1S/C15H15N3O.C3H6O3.H2O/c1-2-19-10-4-6-13-12(8-10)15(17)11-5-3-9(16)7-14(11)18-13;1-2(4)3(5)6;/h3-8H,2,16H2,1H3,(H2,17,18);2,4H,1H3,(H,5,6);1H2

InChI key

NYEPHMYJRNWPLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate is also known as acrinol, is used as a building block in the synthesis of amino acid derivatives and oligonucleotides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuping Hou et al.
European journal of obstetrics, gynecology, and reproductive biology, 151(2), 149-153 (2010-05-04)
To compare the effectiveness and safety of mifepristone/misoprostol versus intra-amniotic injection of ethacridine lactate for the termination of second trimester pregnancy. 210 women requesting voluntary termination of pregnancies at between 16 and 24 weeks of gestation were randomly assigned into
Crystal structure of 6, 9-diamino-2-ethoxyacridinium 3, 5-dinitrobenozate?dimethylsulfoxide?water (1/1/1), C24H27N5O9S
Mirocki, A, et al.
Zeitschrift fur Kristallographie - New Crystal Structures, 237, 639-641 (2022)
Midori Shirato et al.
Journal of photochemistry and photobiology. B, Biology, 173, 434-440 (2017-07-01)
A recently developed antimicrobial technique utilizing hydroxyl radicals generated by hydrogen peroxide (H
K M Minkova et al.
Journal of biotechnology, 102(1), 55-59 (2003-04-02)
C-phycocyanin was purified from Spirulina (Arthrospira) fusiformis by a multi-step treatment of the crude extract with rivanol in a ratio 10:1 (v/v), followed by 40% saturation with ammonium sulfate. After removal of rivanol by gel-filtration on Sephadex G-25, the pigment
Zhiyong Guo et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 856(1-2), 312-317 (2007-08-11)
A HPLC method with UV detection was developed and validated for the simultaneous determination of rivanol and mifepristone in human plasma. Norethisterone was used as the internal standard. Separation was performed by a C18 reversed-phase column maintained at 20 degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service