D150959
1,1-Dimethylbiguanide hydrochloride
97%
Synonym(s):
Metformin
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
NH2C(=NH)NHC(=NH)N(CH3)2 · HCl
CAS Number:
Molecular Weight:
165.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
crystals
mp
223-226 °C (lit.)
SMILES string
Cl[H].CN(C)C(=N)NC(N)=N
InChI
1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H
InChI key
OETHQSJEHLVLGH-UHFFFAOYSA-N
Gene Information
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1,1-Dimethylbiguanide hydrochloride (metformin) is a strong base. It forms well defined salts and possessing excellent coordination capacity with transition metals, forming highly colored bidentate chelate complexes. Metformin (MET) exists in various forms: diprotonated (H2MET)2+ in strong acidic solution, monoprotonated (HMET)+ in weak acid, MET in neutral and deprotonated (MET)- in strong alkali solution. It is an oral antidiabetic drug. Pharmacokinetics of metformin has been studied. Determination of metformin in human plasma has been described by simple HPLC-UV method. Metformin is reported to increase plasma active glucagon-like peptide-1 (GLP-1) in humans.
Metformin is used as an organic base catalyst for the transesterification of coconut oil with methanol.
Metformin is used as an organic base catalyst for the transesterification of coconut oil with methanol.
Application
1,1-Dimethylbiguanide hydrochloride is suitable biguanide agent used in a study to investigate its role in enhancing the secretions of plasma active glucagon-like peptide-1 (GLP-1) levels. It may be used in the synthesis of the following:
- bis(1,1-dimethylbiguanido)copper(II) octahydrate
- bis(1,1-dimethylbiguanido)nickel(II)
- 1,1-dimethylbiguanidium tetrabromothallate(III)
Biochem/physiol Actions
Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity. Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Bis (1,1-dimethylbiguanido) copper (II) octahydrate.
Zhu M, et al.
Acta Crystallographica Section E, Structure Reports Online, 58(5), 217-219 (2002)
1,1-Dimethylbiguanidium tetrabromothallate (III).
He Z, et al.
Acta Crystallographica Section E, Structure Reports Online, 58(11), m647-m649 (2002)
Bis (1, 1-dimethylbiguanido) nickel (II).
Zhu M, et al.
Acta Crystallographica Section E, Structure Reports Online, 58(6), 272-274 (2002)
Nobuyuki Yasuda et al.
Biochemical and biophysical research communications, 298(5), 779-784 (2002-11-07)
Metformin was reported to increase plasma active glucagon-like peptide-1 (GLP-1) in humans. There are two possible mechanisms for this effect: (1) metformin inhibits dipeptidyl peptidase IV (DPPIV), an enzyme degrading GLP-1, and (2) metformin enhances GLP-1 secretion. To elucidate the
Eric P Kusnadi et al.
The EMBO journal, 39(21), e105111-e105111 (2020-09-19)
Elevated ribosome biogenesis in oncogene-driven cancers is commonly targeted by DNA-damaging cytotoxic drugs. Our previous first-in-human trial of CX-5461, a novel, less genotoxic agent that specifically inhibits ribosome biogenesis via suppression of RNA polymerase I (Pol I) transcription, revealed single-agent
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service