Recommended Products
form
solid
SMILES string
Cc1ccc2cnccc2c1
InChI
1S/C10H9N/c1-8-2-3-10-7-11-5-4-9(10)6-8/h2-7H,1H3
InChI key
PPEJLOXOMBAXES-UHFFFAOYSA-N
1 of 4
This Item | A4018 | A9045 | A9414 |
---|---|---|---|
≥250 g/cm2 (1% gel) | ≥200 g/cm2 (1% gel) | ≥200 g/cm2 (1% gel) | ≥200 g/cm2 (1% gel) |
≤0.12 | ≤0.10 | ≤0.1 | ≤0.10 |
200 | 200 | - | 200 |
gel point 26 °C ±2 °C (1.5% gel) | congealing temperature 26-30 °C | congealing temperature 26-30 °C | congealing temperature 26-30 °C |
electrophoresis: suitable | cell culture | mammalian: suitable | cell culture | insect: suitable, cell culture | mammalian: suitable, cell culture | plant: suitable | - |
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service