Skip to Content
Merck
All Photos(1)

Key Documents

CDS007432

Sigma-Aldrich

copper oxychloride

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H6Cl2Cu4O6
CAS Number:
Molecular Weight:
427.13
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

form

solid

reaction suitability

core: copper
reagent type: catalyst

SMILES string

O[Cu]O.O[Cu]O.O[Cu]O.Cl[Cu]Cl

InChI

1S/2ClH.4Cu.6H2O/h2*1H;;;;;6*1H2/q;;4*+2;;;;;;/p-8

InChI key

JNPOSJBMZIQEKM-UHFFFAOYSA-F

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Vázquez-Blanco et al.
Environmental pollution (Barking, Essex : 1987), 257, 113585-113585 (2019-11-23)
Microbial responses to Cu pollution as a function of Cu sources (Cu salts and commercial Cu fungicides) were assessed in a soil using basal soil respiration, and bacterial and fungal community growth, as endpoints. The soil was amended with different
The influence of Miedzian 50, a cupric pesticide, on the internal organs of adult Xenopus laevis. I. Liver and intestine.
A Maryańska-Nadachowska et al.
Folia biologica, 30(1-2), 51-58 (1982-01-01)
Mareeswaran Jeyaraman et al.
BMC plant biology, 18(1), 222-222 (2018-10-12)
Branch canker caused by Macrophoma theicola is a major stem disease of tea plants (Camellia spp.). In tea plantations, this disease causes crop loss and it is one of the major limiting factor for yield stagnation. In very few instances
Pesticide dissipation capacity of an organic biomixture used in the agriculture exposed to copper oxychloride.
G R Tortella et al.
Ecotoxicology and environmental safety, 190, 110121-110121 (2020-01-04)
Alice Agarbati et al.
PloS one, 14(6), e0217385-e0217385 (2019-06-21)
Yeast species that colonize the surface of grape berries at harvest time play an important role during the winemaking process. In this study, the use of culturable microbial techniques permitted a quantitative and qualitative inventory of the different yeast species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service