Skip to Content
Merck
All Photos(1)

Key Documents

CDS001502

Sigma-Aldrich

o-Benzylhydroxylamine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
Molecular Weight:
123.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

form

liquid

SMILES string

NOCc1ccccc1

InChI

1S/C7H9NO/c8-9-6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

XYEOALKITRFCJJ-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Soma et al.
Clinical and experimental nephrology, 24(2), 174-184 (2019-11-27)
To evaluate whether serum N-glycan profile can be used as a diagnostic marker of graft rejection after living-donor kidney transplants (KT). We retrospectively examined 174 KT recipients at five medical centers. N-Glycan levels were analyzed in postoperative serum samples using
Yasushi Yamasaki et al.
Journal of gastroenterology and hepatology, 30(3), 528-534 (2014-08-30)
Serum glycans have been reported to be promising diagnostic markers for many inflammatory diseases and cancers. The aims of this study were to investigate whole glycan expression in patients with non-alcoholic fatty liver diseases and to evaluate the potential use
Yutaka Akimoto et al.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.], 15(4), 432-438 (2015-06-09)
Diagnosing the invasiveness of intraductal papillary mucinous neoplasms (IPMNs) is difficult, especially by blood test. Alterations in serum glycan profiles have been reported for several cancers, but changes in serum glycan profiles have not been investigated in patients with IPMNs.
Takeshi Tomoda et al.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.], 16(1), 44-51 (2016-01-03)
The aims of this study were to determine the change in whole-serum N-glycan profile in autoimmune pancreatitis (AIP) patients and to investigate its clinical utility. We collected serum from 21 AIP patients before any treatment, and from 60 healthy volunteers
Bo Tan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1146, 122120-122120 (2020-05-04)
Accumulation of Immune Responsive Gene 1(IRG1) in macrophage induced by lipopolysaccharide (LPS) and interferon gamma (IFN-γ) leads to production of itaconate by decarboxylation of cis-aconitate. The biology associated with IRG1 and itaconate is not fully understood. A rapid and sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service