Skip to Content
Merck
All Photos(1)

Key Documents

CDS001209

Sigma-Aldrich

n-Octadecyltriethoxysilane

AldrichCPR

Synonym(s):

Triethoxy(octadecyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H52O3Si
CAS Number:
Molecular Weight:
416.75
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

SMILES string

CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC

InChI

1S/C24H52O3Si/c1-5-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-28(25-6-2,26-7-3)27-8-4/h5-24H2,1-4H3

InChI key

FZMJEGJVKFTGMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kookjoo Kim et al.
Talanta, 182, 273-278 (2018-03-05)
A technique for wafer-level detection of organic contaminations via surface-assisted laser desorption/ionization time-of-flight mass spectrometry was developed. To replace the organic matrix in matrix-assisted laser desorption/ionization time-of-flight mass spectrometry, zinc oxide-reduced graphene oxide (ZnO-rGO) hybrid was prepared by a hydrothermal
Chang Liu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(7), 1801169-1801169 (2019-04-17)
The utilization of an inorganic hole-transport layer (HTL) is one of the most effective methods to improve the stability and reduce the cost of perovskite solar cells (PSCs). However, achieving high-quality inorganic HTL films, especially HTL films in n-i-p structures
Ismail Warad et al.
International journal of molecular sciences, 13(5), 6279-6291 (2012-07-04)
Novel hybrid xerogel materials were synthesized by a sol-gel procedure. n-octadecyltriethoxysilane was co-condensed with and without different cross-linkers using Q(0) and T(0) mono-functionalized organosilanes in the presence of n-hexadecylamine with different hydroxyl silica functional groups at the surface. These polymer
Kota Shiba et al.
Scientific reports, 7(1), 3661-3661 (2017-06-18)
Smells are known to be composed of thousands of chemicals with various concentrations, and thus, the extraction of specific information from such a complex system is still challenging. Herein, we report for the first time that the nanomechanical sensing combined
Pei-Ying Lin et al.
Scientific reports, 3, 1703-1703 (2013-04-24)
Intravital fluorescence imaging has great potential in biological and biomedical research, as it provides the ability to directly observe biological structures and processes in their natural state. Contrast agents for intravital imaging applications should exhibit good biocompatibility, multiphoton fluorescence, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service