Skip to Content
Merck
All Photos(1)

Key Documents

CDS000039

Sigma-Aldrich

3-Methyl-2-oxopentanoic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
Molecular Weight:
130.14
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

solid

SMILES string

CC(C(C(O)=O)=O)CC

InChI

1S/C6H10O3/c1-3-4(2)5(7)6(8)9/h4H,3H2,1-2H3,(H,8,9)

InChI key

JVQYSWDUAOAHFM-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Oldford et al.
Free radical biology & medicine, 135, 15-27 (2019-02-23)
Here, we examined the hydrogen peroxide (H2O2) producing capacities of pyruvate dehydrogenase (PDH), α-ketoglutarate dehydrogenase (KGDH), proline dehydrogenase (PRODH), glycerol-3-phosphate dehydrogenase (G3PDH), succinate dehydrogenase (SDH; complex II), and branched-chain keto acid dehydrogenase (BCKDH), in cardiac and liver mitochondria isolated from
Lixia Chen et al.
Journal of food science, 82(10), 2269-2275 (2017-09-08)
Phenyllactic acid (PLA) is a novel antimicrobial compound. A novel NADH-dependent d-lactate dehydrogenase (d-LDH), named as LF-d-LDH0653, with high phenylpyruvate (PPA) reducing activity was isolated from Lactobacillus fermentum JN248. Its optimum pH and temperature were 8.0 and 50 °C, respectively.
Nidhi Kuksal et al.
Biochemical and biophysical research communications, 498(1), 214-220 (2018-03-05)
Recent work has found that complex I is the sole source of reactive oxygen species (ROS) during myocardial ischemia-reperfusion (IR) injury. However, it has also been reported that heart mitochondria can also generate ROS from other sources in the respiratory
null
Kelsey H Fisher-Wellman et al.
Cell reports, 24(13), 3593-3606 (2018-09-27)
Chronic metabolic diseases have been linked to molecular signatures of mitochondrial dysfunction. Nonetheless, molecular remodeling of the transcriptome, proteome, and/or metabolome does not necessarily translate to functional consequences that confer physiologic phenotypes. The work here aims to bridge the gap

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service