Skip to Content
Merck
All Photos(1)

Key Documents

CDS000031

Sigma-Aldrich

Oxanthrene

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O2
CAS Number:
Molecular Weight:
184.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

O1c2ccccc2Oc3ccccc13

InChI

1S/C12H8O2/c1-2-6-10-9(5-1)13-11-7-3-4-8-12(11)14-10/h1-8H

InChI key

NFBOHOGPQUYFRF-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiafeng Ding et al.
Journal of hazardous materials, 355, 74-81 (2018-05-19)
Polychlorinated dibenzo-p-dioxins (PCDDs), as a group of notorious anthropogenic environmental toxicants, are arguably ubiquitous in nature. In this study, we investigated the photocatalytic reductive dechlorination of 2-chlorodibenzo-p-dioxin (2-CDD) over Pd/g-C3N4 catalysts under UV-vis irradiation. The g-C3N4 and a series of
Ali Esrafili et al.
Mikrochimica acta, 187(1), 39-39 (2019-12-12)
A novel metal-organic framework called MFU-4 l was synthesized from ZnCl2 and 1H-1,2,3-triazolo[4,5-b][4',5'-i])dibenzo[1,4]dioxin. MFU-4 l was characterized and is shown to be a viable sorbent for spin-column micro-solid phase extraction of 4-chlorophenol, 2,3-dichlorophenol, 2,4-dichlorophenol, and 2,4,6-trichlorophenol. Following extraction and elution with methanol
Eun-Young Kim et al.
Ecotoxicology and environmental safety, 171, 99-111 (2019-01-01)
To understand the role of aryl hydrocarbon receptor (AHR) isoforms in avian species, we investigated the functional characteristics of two AHR isoforms (designated as jcAHR1 and jcAHR2) of the jungle crow (Corvus macrorhynchos). Two amino acid residues corresponding to Ile324
Victoria B F Custodis et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(36), 8658-8668 (2017-04-08)
One of the key challenges in renewable chemical production is the conversion of lignin, especially by fast pyrolysis. The complexity of the lignin pyrolysis process has hindered the elucidation of the mechanism, inhibiting further industrial implementation. By combining pyrolysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service