Skip to Content
Merck
All Photos(1)

Key Documents

CDS000024

Sigma-Aldrich

2-(2-Aminoethoxy)ethylamine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12N2O
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

form

liquid

SMILES string

NCCOCCN

InChI

1S/C4H12N2O/c5-1-3-7-4-2-6/h1-6H2

InChI key

GXVUZYLYWKWJIM-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oscar Hernandez et al.
Scientific reports, 8(1), 13768-13768 (2018-09-15)
Optogenetics provides tools to control afferent activity in brain microcircuits. However, this requires optical methods that can evoke asynchronous and coordinated activity within neuronal ensembles in a spatio-temporally precise way. Here we describe a light patterning method, which combines MHz
Zhili Xu et al.
Pharmacognosy magazine, 13(51), 454-458 (2017-08-26)
Berberine is a natural isoquinoline alkaloid possessing various pharmacological effects, particularly apparent in the treatment of diarrhea, but the underlying mechanism remains unclear. Smooth muscle myosin light-chain kinase (MLCK) plays a crucial role in the smooth muscle relaxation-contraction events, and
Alamgeer et al.
Journal of ethnopharmacology, 225, 211-219 (2018-07-17)
Thymus linearis, Benth indigenous to Pakistan has been traditionally used for the treatment of various diseases including hypertension. Present study aims to investigate vasorelaxant effect of Thymus linearis and its underlying vasorelaxation mechanisms in porcine coronary artery rings. Aqueous-methanolic extract
Taseer Ahmad et al.
Clinical and experimental hypertension (New York, N.Y. : 1993), 42(4), 335-344 (2019-09-17)
Background: Juglone, a natural phenolic compound obtained from the walnut tree, is known for its wide range of biological activities. However, it has yet to be tested for its effects on hypertension and vascular tone. This investigation was aimed to
Mohammad Mehdi Ommati et al.
Nutritional neuroscience, 23(9), 731-743 (2019-03-12)
Objective(s): Manganese (Mn) is an essential trace element physiologically incorporated in the structure of several vital enzymes. Despite its essentiality, excessive Mn exposure is toxic with brain tissue as the primary target organ. There is no specific and clinically available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service