Skip to Content
Merck
All Photos(1)

Documents

C59122

Sigma-Aldrich

1-Chloro-4-nitrobenzene

99%

Synonym(s):

1-Nitro-4-chlorobenzene, 4-Chloro-1-nitrobenzene, 4-Chloronitrobenzene, 4-Nitro-1-chlorobenzene, PNCB, p-Nitrochlorobenzene, p-Nitrophenyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
508691
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.4 (vs air)

vapor pressure

0.09 mmHg ( 25 °C)

Assay

99%

form

crystals

autoignition temp.

500 °F

bp

242 °C (lit.)

mp

80-83 °C (lit.)

density

1.298 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Cl)cc1

InChI

1S/C6H4ClNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H

InChI key

CZGCEKJOLUNIFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Muta. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qiang Chen et al.
Chemistry, an Asian journal (2018-05-01)
Platinum nanoparticles encapsulated into zeolite Y (Pt@Y catalyst) exhibit excellent catalytic selectivity in the hydrogenation of substituted nitroarenes to form the corresponding aromatic amines, even after complete conversion. With the hydrogenation of p-chloronitrobenzene as a model, the role of zeolite encapsulation
Ruiyang Qu et al.
Nanomaterials (Basel, Switzerland), 8(9) (2018-09-08)
The solvent-free selective hydrogenation of nitrobenzene was carried out using a supported AuPd nanoparticles catalyst, prepared by the modified impregnation method (MIm), as efficient catalyst >99% yield of aniline (AN) was obtained after 15 h at 90 °C, 3 bar
Liang Zhu et al.
Water research, 46(19), 6291-6299 (2012-10-17)
Using a combined zero-valent iron (ZVI) and anaerobic sludge system as the platform, the hydrogen utilization and reductive mechanism of p-chloronitrobenzene (p-ClNB) were investigated in the study. Results showed that the corrosion of ZVI could be used as a source
Yun Zhang et al.
Biodegradation, 20(1), 55-66 (2008-05-30)
Comamonas sp. strain CNB-1 can utilize 4-chloronitrobenzene (4CNB) as sole carbon and nitrogen source for growth. Previous studies were focused on 4CNB degradative pathway and have showed that CNB-1 contained a plasmid pCNB1 harboring the genes (cnbABCaCbDEFGH, cnbZ) for the
Miaomiao Ye et al.
Journal of hazardous materials, 184(1-3), 612-619 (2010-09-22)
TiO(2) hollow microspheres were synthesized by a simple hydrothermal method followed by calcination at different temperatures ranging from 400 to 800°C. The prepared samples were characterized by XRD, SEM, TEM, SAED, HRTEM, N(2) adsorption, and UV-vis spectroscopy. The photocatalytic activities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service