Skip to Content
Merck
All Photos(2)

Key Documents

C58002

Sigma-Aldrich

6-Chloronicotinamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClN2O
CAS Number:
Molecular Weight:
156.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

210-212 °C (lit.)

SMILES string

NC(=O)c1ccc(Cl)nc1

InChI

1S/C6H5ClN2O/c7-5-2-1-4(3-9-5)6(8)10/h1-3H,(H2,8,10)

InChI key

ZIJAZUBWHAZHPL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Fabiano et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 130, 281-289 (2018-07-15)
Nanoparticles (NP) only different in mucoadhesivity are compared for impact on drug oral bioavailability. Two polymeric NP types based on quaternary ammonium-chitosan (NP QA-Ch) and S-protected thiolated derivative thereof (NP QA-Ch-S-pro), respectively, containing the macromolecular drug model, FD4, were prepared
Heather S Loring et al.
Biochemistry, 57(38), 5524-5532 (2018-08-28)
Nicotinamide N-methyltransferase (NNMT) catalyzes the transfer of a methyl group from S-adenosylmethionine (SAM) to nicotinamide, pyridine, and other structural analogues. Aberrantly increased NNMT activity results in the depletion of SAM, nicotinamide (NAM), and nicotinamide adenine dinucleotide (NAD+); NAM is required
Noemi Lupo et al.
Acta biomaterialia, 64, 106-115 (2017-10-17)
Synthesis and evaluation of an entirely S-protected chitosan as mucoadhesive excipient for vaginal drug delivery. N-acetyl-cysteine was linked to 6-mercaptonicotinamide via disulphide exchange reaction. The obtained ligand, NAC-6-MNA, was subsequently attached to chitosan by carbodiimide mediated amide bond formation in
Kesinee Netsomboon et al.
Carbohydrate polymers, 242, 116395-116395 (2020-06-23)
The potential of Cys-Cys ligands for the development of a novel type of S-protected thiomers was evaluated. S-protected thiomers chitosan-N-acetylcysteine-mercaptonicotinamide (CS-NAC-MNA) and chitosan-N-acetylcysteine-N-acetylcysteine (CS-NAC-NAC) were synthesized and characterized. Viscosity of polymers in presence of various concentrations of S-amino acids was
Harshini Neelakantan et al.
Biochemical pharmacology, 147, 141-152 (2017-11-21)
There is a critical need for new mechanism-of-action drugs that reduce the burden of obesity and associated chronic metabolic comorbidities. A potentially novel target to treat obesity and type 2 diabetes is nicotinamide-N-methyltransferase (NNMT), a cytosolic enzyme with newly identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service