Skip to Content
Merck
All Photos(1)

Key Documents

BR781662

BRAND® microplate BRANDplates®, pureGradeS

size 96 wells, size 350 μL, flat bottom, transparent

Synonym(s):

BRAND® microplate BRANDplates®, pureGrade, BRANDplates®, Microplates, pureGradeS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

colorless wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)
with lid

packaging

pack of 1 ea
pkg of (individually packaged)

manufacturer/tradename

BRAND 781662

size

350 μL
96 wells

working volume

350 μL

color

transparent

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • BRANDplates®, pureGradeS microplates are manufactured by high-quality crystal clear polystyrene.
  • Non-treated plates are sterilized by β-radiation for optimal results.
  • Ideal for use in bacteriological assays.
  • Alphanumeric codes facilitate accurate sample identification.
  • The microplates are free from endotoxins, DNase, DNA, RNase, non-cytotoxic.

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service