Skip to Content
Merck
All Photos(1)

Key Documents

BR781621

BRAND® microplate BRANDplates®, pureGrade

size 384 wells, size 100 μL, flat bottom, white

Synonym(s):

BRANDplates®, Microplates, medium binding, pureGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
flat bottom
polystyrene
white wells
size:
100 μL
384 wells
working volume:
100 μL

material

flat bottom
polystyrene
white wells

sterility

non-sterile

feature

lid: no

packaging

pack of 50 ea

manufacturer/tradename

BRAND 781621

size

100 μL
384 wells

working volume

100 μL

color

white

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a medium-binding, untreated surface, making it an ideal choice for a variety of applications. This non-sterile microplate serves as the standard for use in homogeneous assays, screenings, and sample storage. It is non-cytotoxic, making it safe for use in cell-based assays. With BIO-CERT® CERTIFIED QUALITY, this microplate is guaranteed to be free from human DNA, RNase, DNase, and pyrogens. This versatile microplate is a dependable option for researchers looking for consistent performance across numerous laboratory applications.

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service