BR701460
BRAND® Reservoir with cover
polypropylene, capacity 40 mL
All Photos(1)
About This Item
UNSPSC Code:
41121517
NACRES:
NB.01
Recommended Products
material
polypropylene
manufacturer/tradename
BRAND 701460
capacity
40 mL
General description
The BRAND® Reservoir with Cover is designed for both manual pipetting and automation, providing a reliable solution for liquid handling in laboratory settings. Made from durable polypropylene (PP), this reservoir has a working volume of 40 ml and features a V-bottom design that minimizes dead volume, ensuring efficient liquid use. The graduated markings allow for precise measurement, enhancing accuracy in experiments.CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG