Key Documents
BR159600
BRAND® water jet filter pump
polypropylene
Synonym(s):
Water jet filter pump
About This Item
UNSPSC Code:
41105100
NACRES:
NB.79
material
polypropylene
manufacturer/tradename
BRAND 159600
General description
For generating vacuum and extracting liquids and vapors*.
Designed for excellent performance: Very low water consumption, constant ulitmate pressure and high suction capacity.
  • High chemical resistance, fluid path consists of PP, FKM and PTFE
  • Operating temperature up to max. 80°C
  • Integrated non-return valve increases safety
  • Simple operation and easy to clean
  • Detachable vacuum connection
  • Variety of supplied adapters simplify connections to most water sources
  • Optional reducing adapters are available
Includes: Water connection (sleeve nut R 3/4 in., reducing adapter R 1/2 in., and tubing connector (hose nozzle) of 10-12 mm outer-), suction line connection (Detachable hose nozzle of 6-9 mm outer-, with screw cap GL 14).
* If necessary, insert an in-line over-flow vessel, or cold trap.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1